Kimberly-Clark Professional™ WypAll™ X70 Foodservice Wipers
Combine superior performance and strength with guaranteed savings. Recent testing shows that WypAll™ Foodservice Towels made with Hydroknit™ Fast Absorbing Material wipe out some well-known foodservice towels. Sturdy enough for extended use. May be machine washed.
Supplier:  Kimberly-Clark Professional™ 05927
 View more versions of this product
Catalog No. 19-137-200
Explore available promotions
Description
Description
  • 1 case = 1 box, 300 sheets per box
  • WypAll X70 are durable, absorbent cloths that stay strong for cleaning performance; features textured surface to pick up food particles
  • Each industrial cloth measures 12.5 in. x 23.5 in. (unfolded) cleans oil, dirt, grime and solvents; Comes quarterfolded for easy storage
  • The wicking power of HydroKnit™ picks up water almost instantly, and hangs on for fewer drips. Laundered shop rags can’t match that kind of performance.
  • Pound for pound, WYPALL X70 hold more oil than rental shop towels and shop rags; One pop-up box replaces 10 lbs. of rags
Specifications
Specifications
Wypall X70 Foodservice Towel
59.69 cm
31.75 cm
Blue
23.5 x 12.5 in. (59.69 x 31.75 cm)
23.5 in.
12.5 in.
Non-woven Polyproylene Cellulose
Reusable
Ideal for all of your general cleaning need