Kimberly-Clark Professional™ WypAll™ Waterless Cleaning Wipes Refill
75 count refill bag for WypAll Waterless Cleaning Wipes Bucket. Ideal presaturated cleaner for on-the-go cleaning of hands, tools, parts and non-porous work surfaces. Natural solvents cut through grease, grime and oil while soothing ingredients aid in moisturizing the skin.
Supplier:  Kimberly-Clark Professional™ 91367
 View more versions of this product
Catalog No. 19-130-1968
Explore available promotions
Description
Description
  • 6 refill packs of 75 sheets; 450 sheets per case (9.5 in. x 12 in. each)
  • Disposable industrial cleaning wipes are pre-saturated with an alcohol-free natural citrus cleaning agent for the waterless cleansing of skin and hard surfaces
  • These wipers are tough on grease, but gentle enough to use on your hand; contains moisturizers to help keep skin hydrated
  • One textured side for scrubbing dirt, grime, and grease (great for tool cleaning), one smooth side for wiping dirt off surfaces and hands
  • Since these wipes are pre-moistened, you won't worry about spilling a liquid cleansing agent
Specifications
Specifications
Waterless Hand Wipe
Wipe
For cleaning of hands, tools, parts and non-porous work surfaces
450 Pk.
Green
Terpenes and Terpenoids (orange) 0 to 8%, Glycerin (Glycerol) 0 to 3%, non-hazardous ingredients 89 to 100%
Refill, bag