missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ WypAll™ Waterless Cleaning Wipes Refill
75 count refill bag for WypAll Waterless Cleaning Wipes Bucket. Ideal presaturated cleaner for on-the-go cleaning of hands, tools, parts and non-porous work surfaces. Natural solvents cut through grease, grime and oil while soothing ingredients aid in moisturizing the skin.
$78.38 - $126.14
Specifications
Product Type | Waterless Hand Wipe |
---|---|
Color | Green |
Form | Wipe |
Composition | Terpenes and Terpenoids (orange) 0 to 8%, Glycerin (Glycerol) 0 to 3%, non-hazardous ingredients 89 to 100% |
For Use With (Application) | For cleaning of hands, tools, parts and non-porous work surfaces |
Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
19-130-1968
|
Kimberly-Clark Professional™
91367 |
Refill, bag |
Case of 450 Each for $78.38
|
|
|||||
19-130-1967
|
Kimberly-Clark Professional™
91371 |
Bucket |
Case of 450 Each for $126.14
|
|
|||||
Description
- 6 refill packs of 75 sheets; 450 sheets per case (9.5 in. x 12 in. each)
- Disposable industrial cleaning wipes are pre-saturated with an alcohol-free natural citrus cleaning agent for the waterless cleansing of skin and hard surfaces
- These wipers are tough on grease, but gentle enough to use on your hand; contains moisturizers to help keep skin hydrated
- One textured side for scrubbing dirt, grime, and grease (great for tool cleaning), one smooth side for wiping dirt off surfaces and hands
- Since these wipes are pre-moistened, you won't worry about spilling a liquid cleansing agent
Specifications
Waterless Hand Wipe | |
Wipe | |
For cleaning of hands, tools, parts and non-porous work surfaces |
Green | |
Terpenes and Terpenoids (orange) 0 to 8%, Glycerin (Glycerol) 0 to 3%, non-hazardous ingredients 89 to 100% | |
450 Pk. |