Kimberly-Clark Professional™ WypAll™ X70 Wipers in a Bucket
Durable, absorbent wipers that stay strong for better cleaning performance. Cleans oil, grime, and solvents. Portable and weather-resistant bucket for on-the-go professionals. Reusable for long lasting performance. Made with our high-tech HYDROKNIT™ Fast Absorbing Material. Ideal for all manufacturing and maintenance & repair.
Supplier:  Kimberly-Clark Professional™ 83571
 View more versions of this product
Catalog No. 19-168-2146
Explore available promotions
Description
Description
  • Wypall X70 Cloths In a Bucket Refill includes 3 Rolls of 220 cloths (case total = 660 cloths)
  • WypAll X70 are durable, absorbent cloths that stay strong for cleaning performance and the portable bucket helps keep the cloths clean
  • Each industrial cloth measures 9.75 in. x 13 in. and cleans oil, dirt, grime and solvents
  • The wicking power of HydroKnit picks up water almost instantly, and hangs on for fewer drips; Laundered shop rags can't match the performance
  • Pound for pound, WYPALL X70 manufactured rags hold more oil than rental shop towels and shop rags
Specifications
Specifications
Medium-duty Cloth
33 cm
25.4 cm
White
Single-use
Ideal for all manufacturing and maintenance and repair
13 in.
13 in.
Non-woven Polyproylene Cellulose
Refill Pack
13 x 10 in. (33 x 25.4 cm)
Non-sterile
Product Suggestions
Product Suggestions