DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures
Highly resistant to temperature and steam during autoclaving
Supplier: DWK Life Sciences 7380213415
Description
- Cemented-in PTFE-faced 14B white rubber liner
- Three screw-thread finishes
Specifications
Black | |
500 Cs. | |
ASTM E982 | |
Screw Cap | |
PTFE/14B Rubber |
Phenolic Resin | |
13-415 | |
Autoclavable | |
Bottle |