missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

Highly resistant to temperature and steam during autoclaving

$494.15 - $679.53

Specifications

Color Black
Material Phenolic Resin
Autoclavable Autoclavable
Product Type Screw Cap
For Use With (Equipment) Bottle
View More Specs

Products 3
Catalog Number Mfr. No. Neck Finish Price Quantity  
Catalog Number Mfr. No. Neck Finish Price Quantity  
05-569-5 Edge
View Documents Promotion Details
DWK Life Sciences
7380215415
15-415
Case of 500 Each for $543.53
Edge
 
05-569-4
View Documents Promotion Details
DWK Life Sciences
7380213415
13-415
Case of 500 Each for $494.15
 
06-421-20
View Documents Promotion Details
DWK Life Sciences
7380222400
22-400
Case of 144 Each for $679.53
 
Description

Description

  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes

Specifications

Specifications

Black
Autoclavable
Bottle
Phenolic Resin
Screw Cap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'