DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures Edge
Edge
Highly resistant to temperature and steam during autoclaving
Supplier:  DWK Life Sciences 7380215415
 View more versions of this product
Catalog No. 05-569-5
Edge
Explore available promotions
Description
Description
  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes
Specifications
Specifications
Black
PTFE-faced Rubber
Yes
Screw Cap
Phenolic Resin
15-415
Autoclavable
Bottle