DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures
Highly resistant to temperature and steam during autoclaving
Supplier:  DWK Life Sciences 7380222400
 View more versions of this product
Catalog No. 06-421-20
Explore available promotions
Description
Description
  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes
Specifications
Specifications
Black
144 Cs.
Autoclavable
Bottle
Phenolic Resin
22-400
Screw Cap
PTFE/14B Rubber