DWK Life Sciences Kimble™ KIMAX™ Short Stem 58° Angle Addition Funnels
Bowls pressed to exact angle for fast filtering
Supplier:  DWK Life Sciences 2895045
 View more versions of this product
Catalog No. 10-322B
Explore available promotions
Description
Description
Bowls pressed to exact angle for fast filtering
  • Heavy uniform walls
  • Beaded reinforced rims
  • Fire-polished ends
Specifications
Specifications
4.5cm
0.65cm
ASTM E1095, Type I, Class A
7cm
5cm
19 mL
Borosilicate Glass
58°
Bowls pressed to exact angle for fast filtering