DWK Life Sciences Kimble™ KIMAX™ Short Stem 58° Angle Addition Funnels
Bowls pressed to exact angle for fast filtering
$200.09 - $603.65
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity | |||
10-322J
|
DWK Life Sciences
2895025 |
3 mL | 0.6cm | 2.5cm | 4cm |
Case of 24 Each for $603.65
|
|
|||
10-322A
|
DWK Life Sciences
2895035 |
8 mL | 6.5 mm | 35mm | 5 cm |
Pack of 6 Each for $200.09
Case of 4 Pack for $800.36
|
|
|||
10-322B
|
DWK Life Sciences
2895045 |
19 mL | 0.65cm | 4.5cm | 5cm |
Pack of 6 Each for $207.74
Case of 4 Pack for $830.96
|
|
|||
10-322F
|
DWK Life Sciences
2895090 |
150 mL | 0.9cm | 9cm | 9.7cm |
Pack of 6 Each for $279.85
Case of 4 Pack for $1,119.40
|
|
|||
10-322D
|
DWK Life Sciences
2895065 |
60 mL | 0.8cm | 6.5cm | 6.3cm |
Pack of 12 Each for $509.94
Case of 4 Pack for $2,039.76
|
|
|||
10-322C
|
DWK Life Sciences
2895055 |
36 mL | 0.8cm | 5.5cm | 6.3cm |
Pack of 12 Each for $425.15
Case of 4 Pack for $1,700.60
|
|
|||
10-322E
|
DWK Life Sciences
2895075 |
90 mL | 0.8cm | 7.5cm | 7.5cm |
Pack of 12 Each for $550.94
Case of 4 Pack for $2,203.76
|
|
|||
10-322G
|
DWK Life Sciences
28950100 |
225 mL | 0.9cm | 10cm | 9.7cm |
Pack of 6 Each for $356.32
Case of 4 Pack for $1,425.28
|
|
|||
Description
Bowls pressed to exact angle for fast filtering
- Heavy uniform walls
- Beaded reinforced rims
- Fire-polished ends
Specifications
Borosilicate Glass |