DWK Life Sciences Kimble™ KIMAX™ Squibb Separatory Funnels with PTFE-Plug Stopcock
Do not require grease
Supplier:  DWK Life Sciences 29048F2000
 View more versions of this product
Catalog No. 10-437-10F
Description
Description
Do not require grease
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed.
  • Eliminate contamination
  • Will not bind or freeze
Order Info
Stem lengths, 60mm.
Compliance
Designed from ASTM™ E 1096-86.
Specifications
Specifications
2 L
ASTM E1096, Type IV
60mm
6; PTFE
38
Borosilicate Glass
Eliminates contamination
Each
Glass
Squibb Separatory Funnel