DWK Life Sciences Kimble™ KIMAX™ Squibb Separatory Funnels with PTFE-Plug Stopcock
Do not require grease
$346.38 - $1074.24
Specifications
Material | Borosilicate Glass |
---|---|
Certifications/Compliance | ASTM E1096, Type IV |
For Use With (Application) | Eliminates contamination |
Length (Metric) Stem | 60mm |
Stopper Material | Glass |
Catalog Number | Mfr. No. | Capacity (Metric) | Stopcock | Stopper Number | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Stopcock | Stopper Number | Quantity | Price | Quantity | ||
10-437-10E
|
DWK Life Sciences
29048F1000 |
1 L | 4; PTFE | 27 | Each |
Each for $661.26
Case of 2 Each for $1,322.52
|
|
||
10-437-10F
|
DWK Life Sciences
29048F2000 |
2 L | 6; PTFE | 38 | Each |
Each for $1,074.24
Case of 2 Each for $2,148.48
|
|
||
10-437-10B
|
DWK Life Sciences
29048F125 |
125 mL | 2; PTFE | 22 | 2 Pack |
Pack of 2 Each for $346.38
Case of 2 Pack for $692.76
|
|
||
10-437-10A
|
DWK Life Sciences
29048F60 |
60 mL | 2; PTFE | 16 | 2 Pack |
Pack of 2 Each for $728.68
Case of 2 Pack for $1,457.36
|
|
||
10-437-10AA
|
DWK Life Sciences
29048F30 |
30 mL | 2; PTFE | 13 | 2 Pack |
Pack of 2 Each for $571.41
Case of 2 Pack for $1,142.82
|
|
||
10-437-10C
|
DWK Life Sciences
29048F250 |
250 mL | 4; PTFE | 22 | 2 Pack |
Pack of 2 Each for $834.76
Case of 2 Pack for $1,669.52
|
|
||
10-437-10D
|
DWK Life Sciences
29048F500 |
500 mL | 4; PTFE | 27 | Each |
Each for $488.18
Case of 4 Each for $1,952.72
|
|
||
Description
Do not require grease
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed.
- Eliminate contamination
- Will not bind or freeze
Order Info
Stem lengths, 60mm.
Compliance
Designed from ASTM™ E 1096-86.
Specifications
Borosilicate Glass | |
Eliminates contamination | |
Glass |
ASTM E1096, Type IV | |
60mm | |
Squibb Separatory Funnel |