DWK Life Sciences Kimble™ Kontes™ Allihn Condensers with standard taper Joints
Drip tip at end of inner joint
Supplier:  DWK Life Sciences 4310002440
 View more versions of this product
Catalog No. K431000-2440
Explore available promotions
Description
Description
  • 24/40 inner and outer joints
  • Hose connections accept 7.9mm I.D. flexible tubing
Compliance
Designed from Fed. Spec. NNN-C-1504 Type III
Specifications
Specifications
Standard taper drip joint at bottom and a standard taper outer joint at top
24/40
57 cm
22.4 in.
Borosilicate Glass
0.37 in.
15.7 in.
Refluxing operations
Allihn Condenser
40 cm
0.952 cm
USP Type I and ASTM E438 Type I, Class A