DWK Life Sciences Kimble™ Kontes™ Micro Condenser Distillation Columns
Supplied jacket allows column to be used as a condenser
Supplier:  DWK Life Sciences 2868100200
 View more versions of this product
Catalog No. K2868100200
Explore available promotions
Description
Description
  • Outer standard taper joint at top
  • Inner standard taper joint with drip tip at bottom
  • Indentation at bottom supports packing material
  • Accept No. 1 hose connections
Specifications
Specifications
200mm
USP Type I, ASTM E438, Type I, Class A
19/22
Jacketed Distillation Column
Condenser