DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Cyanide Distillation System, Complete
Complete system and component parts for cyanide distillation
Supplier: DWK Life Sciences™ 4794004000
Description
Includes 10 sets of glassware, heater, and pre-cut tubing.
Specifications
10 sets of glassware, heater, and pre-cut tubing | |
MIDI-VAP™ 4000 Cyanide Complete System | |
USP Type I and ASTM E438, Type I, Class A |
Designed to support the semi-automatic colorimetric methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes | |
Borosilicate Glass |