DWK Life Sciences Kimble™ Kontes™ MIDI-VAP 4000 Cyanide Distillation System, Complete
Complete system and component parts for cyanide distillation
$527.18 - $45974.50
Specifications
For Use With (Application) | Designed to support the semi-automatic colorimetric methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes |
---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | |||||
K4794620023
|
DWK Life Sciences
4794620023 |
Absorption Impinger | Coarse porosity frit |
Case for $527.18
|
|
|||||
K4794004000
|
DWK Life Sciences
4794004000 |
MIDI-VAP™ 4000 Cyanide Complete System | 10 sets of glassware, heater, and pre-cut tubing |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Includes 10 sets of glassware, heater, and pre-cut tubing.
Specifications
Designed to support the semi-automatic colorimetric methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes |