DWK Life Sciences Kimble™ Kontes™ MIDI-VAP 4000 Cyanide Distillation System, Complete
Complete system and component parts for cyanide distillation
Supplier: DWK Life Sciences 4794620023
Description
Includes 10 sets of glassware, heater, and pre-cut tubing.
Specifications
Coarse porosity frit | |
Absorption Impinger | |
29/42 |
Designed to support the semi-automatic colorimetric methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes | |
MIDI-VAP™ 4000 cyanide distillation system |