missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ PTFE-Plugged Columns

Manufactured from borosilicate glass 3.3

Supplier:  DWK Life Sciences 4205400234

 View more versions of this product

Catalog No. K420540-0234


Explore available promotions

Description

Description

With PTFE stopcock plug

  • Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
  • Use glass wool to support packing in columns without fritted disc
  • PTFE plug controls flowrate
  • Packing not included

Specifications

Specifications

22 mm
40 to 60 μm
400 mm
Fritted Disc, 2mm Bore PTFE Stopcock Plug
Gravity Chromatography Column
150 mL
Glass
Videos
SDS
Documents

Documents

Promotions

Promotions