DWK Life Sciences Kimble™ Kontes™ PTFE-Plugged Columns
Manufactured from borosilicate glass 3.3
$269.32 - $715.30
Specifications
Material Glass
View More Specs
Products 20
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Column Capacity Includes Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Column Capacity Includes Price Quantity  
K420530-0221
View Documents Promotion Details
DWK Life Sciences
4205300221
200 mm 15 mm 30 mL 2mm Bore PTFE Stopcock Plug
Each for $297.60
K420530-0213
View Documents Promotion Details
DWK Life Sciences
4205300213
250 mm 11 mm 20 mL 2mm Bore PTFE Stopcock Plug
Each for $269.32
K420530-0211
View Documents Promotion Details
DWK Life Sciences
4205300211
150 mm 11 mm 10 mL 2mm Bore PTFE Stopcock Plug
Each for $304.58
K420530-0222
View Documents Promotion Details
DWK Life Sciences
4205300222
250 mm 15 mm 40 mL 2mm Bore PTFE Stopcock Plug
Each for $299.08
K420530-0225
View Documents Promotion Details
DWK Life Sciences
4205300225
500 mm 15 mm 80 mL 2mm Bore PTFE Stopcock Plug
Each for $394.70
K420530-0234
View Documents Promotion Details
DWK Life Sciences
4205300234
500 mm 19 mm 140 mL 2mm Bore PTFE Stopcock Plug
Each for $365.81
K420530-0244
View Documents
DWK Life Sciences
4205300244
500 mm 22 mm 190 mL 2mm Bore PTFE Stopcock Plug
Each for $388.13
K420530-0232
View Documents Promotion Details
DWK Life Sciences
4205300232
300 mm 19 mm 80 mL 2mm Bore PTFE Stopcock Plug
Each for $322.26
K420530-0251
View Documents Promotion Details
DWK Life Sciences
4205300251
350 mm 30 mm 220 mL 2mm Bore PTFE Stopcock Plug
Each for $374.26
K420530-0252
View Documents Promotion Details
DWK Life Sciences
4205300252
400 mm 30 mm 250 mL 2mm Bore PTFE Stopcock Plug
Each for $396.38
K420530-0242
View Documents Promotion Details
DWK Life Sciences
4205300242
350 mm 22 mm 130 mL 2mm Bore PTFE Stopcock Plug
Each for $358.77
K420530-0253
View Documents Promotion Details
DWK Life Sciences
4205300253
500 mm 30 mm 300 mL 2mm Bore PTFE Stopcock Plug
Each for $415.89
K420540-0213
View Documents Promotion Details
DWK Life Sciences
4205400213
300 mm 11 mm 20 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $346.49
K420540-0224
View Documents Promotion Details
DWK Life Sciences
4205400224
400 mm 19 mm 110 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $371.72
K420530-0415
View Documents
DWK Life Sciences
4205300415
700 mm 49 mm 1300 mL 2mm Bore PTFE Stopcock Plug
Each for $715.30
K420540-0233
View Documents Promotion Details
DWK Life Sciences
4205400233
300 mm 22 mm 110 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $381.13
K420540-0234
View Documents Promotion Details
DWK Life Sciences
4205400234
400 mm 22 mm 150 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $384.74
K420540-0243
View Documents Promotion Details
DWK Life Sciences
4205400243
300 mm 25 mm 140 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $405.83
K420540-0245
View Documents Promotion Details
DWK Life Sciences
4205400245
500 mm 25 mm 240 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $540.45
K420540-0256
View Documents
DWK Life Sciences
4205400256
600 mm 41 mm 775 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug
Each for $597.45
Description
Description
With PTFE stopcock plug
  • Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
  • Use glass wool to support packing in columns without fritted disc
  • PTFE plug controls flowrate
  • Packing not included
Specifications
Specifications
Glass
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'