DWK Life Sciences Kimble™ Kontes™ Vacuum/Gas Double Manifold with Glass Plugs
All-glass manifolds for connecting inert gas and vacuum sources
Supplier: DWK Life Sciences 2161300004
Description
All-glass manifolds for connecting inert gas and vacuum sources
- Fabricated from 0.5 in. medium-wall tubing
- 4mm bore glass stopcocks with lower vacuum chamber spaced 85mm apart
- Top right connection is for inert gas
- Lower left connection is for vacuum
Specifications
Double Manifold | |
Glass | |
0.5 in. | |
14.3 in. |
0.5 in. Medium wall tubing, stopcocks, 4mm bore Glass, lower vacuum chamber, 2 hose connections | |
4 | |
1.27 cm | |
36.5 cm |