DWK Life Sciences Kimble™ Kontes™ Vacuum/Gas Double Manifold with Glass Plugs
All-glass manifolds for connecting inert gas and vacuum sources
Supplier:  DWK Life Sciences 2161300005
 View more versions of this product
Catalog No. K2161300005
Explore available promotions
Description
Description
All-glass manifolds for connecting inert gas and vacuum sources
  • Fabricated from 0.5 in. medium-wall tubing
  • 4mm bore glass stopcocks with lower vacuum chamber spaced 85mm apart
  • Top right connection is for inert gas
  • Lower left connection is for vacuum
Specifications
Specifications
Double Manifold
Glass
0.5 in.
17.7 in.
0.5 in. Medium wall tubing, stopcocks, 4mm bore Glass, lower vacuum chamber, 2 hose connections
5
1.27 cm
45 cm