DWK Life Sciences Kimble™ Micro Hematocrit Capillary Tubes Edge
Edge
Kimble™ micro-hematocrit tubes are used to measure the volume percentage of red blood cells in blood.
Supplier:  DWK Life Sciences 41A2502
 View more versions of this product
Catalog No. 22-037-520
Edge
Explore available promotions
Description
Description
Kimble™ micro-hematocrit tubes are used to measure the volume percentage of red blood cells in blood.
  • Packed in plastic vials with reclosable snap-lock caps to maintain cleanliness
  • Ends may be sealed by flame or sealing clay
  • Heparinized capillary tubes are designed to prevent blood clotting
  • Ammonium heparin does not interfere with sodium identification
  • Tubes are manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
Specifications
Specifications
Soda Lime Glass
1.1 mm
75 mm
60 mm
Snap-lock
Used to measure the volume percentage of red Blood cells in Blood
USP type III