DWK Life Sciences Kimble Chase™ Short Form High Precision Specific Gravity Hydrometers
Available in a variety of graduation ranges and overall lengths
Supplier:  DWK Life Sciences 521120910
 View more versions of this product
Catalog No. 13-757-061
Explore available promotions
Description
Description
  • Serial number is printed on the scale
  • Ballast is made from steel pellets and a binder, and is free of heavier metals
  • Includes Certificate of Traceability
  • Certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
  • Meets or exceeds the requirements in ASTM™ specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
Specifications
Specifications
Short Form High Precision Specific Gravity Hydrometer
165 mm
5 mm
0.940 to 1.010
0.001 sp.gr.
Certificate of Traceability
18 mm
140 mL