missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Clear Glass Straight-Sided Jars, Tall: Pulp/Vinyl Cap

Wide-mouth design

$141.88 - $230.00

Specifications

Color Clear
For Use With (Application) Storage
Includes Cap
Material Soda Lime Glass
Closure Color Black
View More Specs

Products 8
Catalog Number Mfr. No. Capacity Closure Size Diameter (Metric) Packaging Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Diameter (Metric) Packaging Price Quantity  
13-774-44
View Documents Promotion Details
DWK Life Sciences
5510448V81
4 oz., 125 mL 48-400 mm 51 mm Convenience
Case of 24 Each for $156.18
 
13-774-48
View Documents Promotion Details
DWK Life Sciences
5510448C81
4 oz., 125 mL 48-400 mm 51 mm Bulk
Case of 24 Each for $230.00
 
13-774-49
View Documents Promotion Details
DWK Life Sciences
5510858C81
8 oz., 250 mL 58-400 mm 62 mm Bulk
Case of 24 Each for $199.82
 
13-774-45
View Documents Promotion Details
DWK Life Sciences
5510858V81
8 oz., 250 mL 58-400 mm 62 mm Convenience
Case of 24 Each for $158.97
 
13-774-50
View Documents Promotion Details
DWK Life Sciences
5511670C81
16 oz., 473 mL 70-400 mm 76 mm Bulk
Case of 12 Each for $173.09
 
13-774-46
View Documents Promotion Details
DWK Life Sciences
5511670V81
16 oz., 473 mL 70-400 mm 76 mm Convenience
Case of 12 Each for $177.91
 
13-774-51
View Documents Promotion Details
DWK Life Sciences
5513289C81
32 oz., 1000 mL 89-400 mm 95 mm Bulk
Case of 12 Each for $141.88
 
13-774-47
View Documents Promotion Details
DWK Life Sciences
5513289V81
32 oz., 1000 mL 89-400 mm 95 mm Convenience
Case of 12 Each for $156.09
 
Description

Description

Wide-mouth design

  • Clear glass for easy viewing of contents
  • Type III soda-lime glass
  • Caps are black pulp/vinyl
  • Choose Convenience Packs (caps attached) or Bulk Packs (caps in bags)

All 32-ounce jars in this product family are packed in corrugated cartons with divider cells.

Specifications

Specifications

Clear
Cap
Black
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Pulp/Vinyl
Storage
Soda Lime Glass
Clear
Wide
Straight-sided Jar
Phenolic
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'