DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 15 mL Capacity
Withstand centrifugation up to 2980 RCF
Supplier:  DWK Life Sciences 4515315
 View more versions of this product
Catalog No. 05-537-6A
Explore available promotions
Description
Description
Withstand centrifugation up to 2980 RCF
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
Specifications
Specifications
To contain
Glass
Reusable
0 to 0.2mL in ±0.05mL, >0.2 to 1mL in ±0.1mL, >1 to 3mL in ±0.15mL, >3mL in ±0.2mL
136 mm
Pennyhead Stopper
17 mm
Centrifuge Tube
USP, ASTM E 438
15 mL
Borosilicate Glass
Conical
Conical
Marking Spot
0.1 mL
2980 x g