DWK Life Sciences Kimble™ Disposable Drum Sampler
Borosilicate glass, open-end drum thieves break easily for quick disposal. Available in 150mL, 25mL and 75mL capacity.
Supplier: DWK Life Sciences 76000150
Description
- Conventional open-end drum thieves provide a quick method for sampling unmarked drums
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Drum Sampler | |
5.07 oz. | |
Single-use | |
0.7 in. | |
43 in. |
Borosilicate Glass | |
150 mL | |
Provides a quick method for sampling unmarked drums | |
1.8 cm | |
109.2 cm |