DWK Life Sciences Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End
Without marking spot
Supplier:  DWK Life Sciences 4506016150
 View more versions of this product
Catalog No. 14-925J
Description
Description
Without marking spot
  • Culture tubes made from KG-33 glass feature fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance
  • Designed from ASTM™ specification E982, Type IV requirements
Specifications
Specifications
ASTM E982, Type IV
24 mL
Borosilicate Glass
16 mm
Plain
150 mm
None
Reusable Glass Tube