DWK Life Sciences Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End
Without marking spot
$24.75 - $286.53
Specifications
Certifications/Compliance | ASTM E982, Type IV |
---|---|
Design Type | Plain |
Material | Borosilicate Glass |
Label Type | None |
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
14-925B
|
DWK Life Sciences
45060650 |
0.7 mL | 6 mm | 50 mm |
Pack of 72 Each for $66.35
Case of 10 Pack for $663.50
|
|
|||
14-925J
|
DWK Life Sciences
4506016150 |
24 mL | 16 mm | 150 mm |
Pack of 72 Each for $153.76
Case of 8 Pack for $1,230.08
|
|
|||
14-925E
|
DWK Life Sciences
4506013100 |
10 mL | 13 mm | 100 mm |
Pack of 72 Each for $24.75
Case of 10 Pack for $247.50
|
|
|||
14-925M
|
DWK Life Sciences
4506025150 |
55 mL | 25 mm | 150 mm |
Pack of 72 Each for $286.53
Case of 4 Pack for $1,146.12
|
|
|||
14-925N
|
DWK Life Sciences
4506025200 |
75 mL | 25 mm | 200 mm |
Pack of 24 Each for $104.85
Case of 8 Pack for $838.80
|
|
|||
14-925K
|
DWK Life Sciences
4506019150 |
30 mL | 19 mm | 150 mm |
Pack of 72 Each for $155.00
Case of 8 Pack for $1,240.00
|
|
|||
Description
Without marking spot
- Culture tubes made from KG-33 glass feature fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance
- Designed from ASTM™ specification E982, Type IV requirements
Specifications
ASTM E982, Type IV | |
Plain | |
Borosilicate Glass | |
None |