DWK Life Sciences Kimble™ Phenolic Caps with Tinfoil Liners
Has good resistance to alcohols, hydrocarbons, ketones, oils; not recommended for acids and alkalis.
Supplier:  DWK Life Sciences 75203G28400
 View more versions of this product
Catalog No. 13-757-144
Explore available promotions
Alert:
Not recommended for acids and alkalis.
Description
Description
  • Economical general purpose phenolic screw cap with a tinfoil liner
  • Color: black
  • Best suited for environmental sampling applications
  • Foil is laminated to paper and bonded to a 0.035-in. pulpboard
Specifications
Specifications
Black
144 Cs.
Phenolic Cap
Tin
Phenolic Resin
28-400
Bottle