missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Phenolic Caps with Tinfoil Liners
Has good resistance to alcohols, hydrocarbons, ketones, oils; not recommended for acids and alkalis.
$92.79 - $541.15
Specifications
Color | Black |
---|---|
Material | Phenolic Resin |
No. per Case | 144 Cs. |
Product Type | Phenolic Cap |
For Use With (Equipment) | Bottle |
Alert:
Not recommended for acids and alkalis.
Catalog Number | Mfr. No. | Neck Finish | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | Price | Quantity | |||||
13-757-144
|
DWK Life Sciences
75203G28400 |
28-400 |
Case of 144 Each for $127.71
|
|
|||||
13-757-143
|
DWK Life Sciences
75203G24400 |
24-400 |
Case of 144 Each for $123.85
|
|
|||||
13-757-141
|
DWK Life Sciences
75203G20400 |
20-400 |
Case of 144 Each for $92.79
|
|
|||||
13-757-142
|
DWK Life Sciences
75203G22400 |
22-400 |
Case of 144 Each for $116.03
|
|
|||||
13-757-145
|
DWK Life Sciences
75203G33400 |
33-400 |
Case of 144 Each for $150.62
|
|
|||||
13-757-146
|
DWK Life Sciences
75203G38400 |
38-400 |
Case of 144 Each for $227.59
|
|
|||||
13-757-148
|
DWK Life Sciences
75203G45400 |
45-400 |
Case of 144 Each for $208.90
|
|
|||||
13-757-152
|
DWK Life Sciences
75203G63400 |
63-400 |
Case of 144 Each for $400.32
|
|
|||||
13-757-150
|
DWK Life Sciences
75203G53400 |
53-400 |
Case of 144 Each for $247.56
|
|
|||||
13-757-149
|
DWK Life Sciences
75203G48400 |
48-400 |
Case of 144 Each for $222.10
|
|
|||||
13-757-151
|
DWK Life Sciences
75203G58400 |
58-400 |
Case of 144 Each for $310.67
|
|
|||||
13-757-154
|
DWK Life Sciences
75203G89400 |
89-400 |
Case of 144 Each for $541.15
|
|
|||||
13-757-153
|
DWK Life Sciences
75203G70400 |
70-400 |
Case of 144 Each for $447.41
|
|
|||||
Description
- Low cost
- Environmental sampling
- General purpose
- Tinfoil lining provides good resistance to alcohols, hydrocarbons, ketones, oils
All caps/liners should be tested to ensure leak and compatibility performance with contents.
Specifications
Black | |
144 Cs. | |
Bottle |
Phenolic Resin | |
Phenolic Cap | |
Tin |