DWK Life Sciences Kimble™ Phenolic Caps with Tinfoil Liners
Has good resistance to alcohols, hydrocarbons, ketones, oils; not recommended for acids and alkalis.
Supplier: DWK Life Sciences 75203G38400
Alert:
Not recommended for acids and alkalis.
Description
- Economical general purpose phenolic screw cap with a tinfoil liner
- Color: black
- Best suited for environmental sampling applications
- Foil is laminated to paper and bonded to a 0.035-in. pulpboard
Specifications
Black | |
144 Cs. | |
Phenolic Cap | |
Tin |
Phenolic Resin | |
38-400 | |
Bottle |