DWK Life Sciences Kimble™ Sample Vials Screw Thread KG-33 Borosilicate Glass
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
Supplier:  DWK Life Sciences 60940D6
 View more versions of this product
Catalog No. 0334047
Explore available promotions
Description
Description
  • Glass sample vial includes unattached rubber-lined black phenolic closure
  • Vial top has a screw thread finish
  • Kimax KG-33 borosilicate glass - meets the requirements for ASTM Type 1, Class A and USP Type 1 Glass
  • High chemical resistance
Specifications
Specifications
A
Clear
Borosilicate Glass
Flat
6.5 dr.
Screw Cap (Closed-top)
ASTM Type I, Class A
Rubber
23 mm
24 mL
Screw
Phenolic
85 mm