missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimtech™ A5 Sterile Integrated Hood and Mask

Cleanroom sterile hood with a mask integrated into the hood.

$633.34 - $766.18

Specifications

Product Type Integrated Hood and Mask
Material Polypropylene SMS Fabric
Color White
Closure Type Ties
Disposable Single-use
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-986-053
View Documents
Kimberly-Clark Professional™
47677
X-Large
Case for $633.34
 
19-048-606
View Documents
Kimberly-Clark Professional™
36072
Universal
Case of 75 Each for $766.18
 
Description

Description

Cleanroom sterile hood with a mask integrated into the hood.

  • Average bacterial filtration efficiency (BFE) of 96%
  • Polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-don ties for easy gowning and are folded inside out for aseptic donning
  • Protective hood-and-mask-combos are made without latex, silicone or the BHT preservative
  • Designed for single-use
  • CE certified, Category III
  • Each individually bagged mask and hood has a sterility indicator
  • Manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications

Specifications

Integrated Hood and Mask
White
Single-use
Integrated
Polypropylene SMS Fabric
Ties
Sterile
Class 100 (ISO 5)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'