Kimberly-Clark Professional™ Kimtech™ A6 Liquid Splash Protection Hooded Coveralls
Cleanroom hooded coveralls provide liquid barrier protection with a breathable back panel for use in ISO 7 and 8 environments.
Supplier:  Kimberly-Clark Professional™ 47683
 View more versions of this product
Catalog No. 19-169-223C
Explore available promotions
Description
Description
  • Presented unzipped with the inside of the garment first
  • Arms telescope folded
  • Legs telescope folded with built-in snaps
  • Elastic cuffs and thumb loops
  • No label on the outer surface
  • Hood, front panel, arms and leg made of high performance laminated SMS fabric
  • Back panel made of ultralight weight breathable SMS
  • 1 large bag of 25 coveralls individually packed per case
Specifications
Specifications
Cleanroom Coveralls
SMS
Elastic
Non-sterile
Controlled Environments
Large
Single-use
Hooded
PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection; EN 14126:2003 certified protection against infective agent penetration
Class 10,000-100,000 (ISO 7-8)