Kimberly-Clark Professional™ Kimtech™ A6 Liquid Splash Protection Hooded Coveralls
Cleanroom hooded coveralls provide liquid barrier protection with a breathable back panel for use in ISO 7 and 8 environments.
$229.86 - $294.98
Specifications
Product Type Cleanroom Coveralls
Material SMS
Disposable Single-use
Cuff Style Elastic
Neck Style Hooded
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-169-223A
View Documents
Kimberly-Clark Professional™
47681
Small
Case of 25 Each for $229.86
19-169-223B
View Documents
Kimberly-Clark Professional™
47682
Medium
Case of 25 Each for $233.35
19-169-223C
View Documents
Kimberly-Clark Professional™
47683
Large
Case of 25 Each for $246.53
19-169-223D
View Documents
Kimberly-Clark Professional™
47684
X-Large
Case of 25 Each for $255.06
19-169-223E
View Documents
Kimberly-Clark Professional™
47685
2X-Large
Case of 25 Each for $260.48
19-169-223F
View Documents
Kimberly-Clark Professional™
47686
3X-Large
Case of 25 Each for $284.91
19-169-223G
View Documents
Kimberly-Clark Professional™
47687
4X-Large
Case of 25 Each for $294.98
Description
Description
  • Presented unzipped with the inside of the garment first
  • Arms telescope folded
  • Legs telescope folded with built-in snaps
  • Elastic cuffs and thumb loops
  • No label on the outer surface
  • Hood, front panel, arms and leg made of high performance laminated SMS fabric
  • Back panel made of ultralight weight breathable SMS
  • 1 large bag of 25 coveralls individually packed per case
Specifications
Specifications
Cleanroom Coveralls
Single-use
Hooded
PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection; EN 14126:2003 certified protection against infective agent penetration
Class 10,000-100,000 (ISO 7-8)
SMS
Elastic
Non-sterile
Controlled Environments
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'