Learn More
Thermo Scientific™ KingFisher Apex Purification Systems
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
$116265.00 - $124387.31
Specifications
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate |
---|---|
Plates per Deck | 8 |
Collection Efficiency of Particles | >98% with 2.8 μm particle size |
For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics |
Heating Temperature | +4°C above ambient temperature up to +100°C |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
5400910
|
Thermo Scientific™
5400910 |
KingFisher Apex with 96 PCR head |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
A46941PKG
|
Thermo Scientific™
A46941 |
KingFisher Apex with 96 PCR head, Extended Warranty Package |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
5400920
|
Thermo Scientific™
5400920 |
KIngFisher Apex with 96 Combi head |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
A46943PKG
|
Thermo Scientific™
A46943 |
KingFisher Apex with 96 Combi head, Extended Warranty Package |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
5400930
|
Thermo Scientific™
5400930 |
KingFisher Apex with 96 DW head |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
A46944PKG
|
Thermo Scientific™
A46944 |
KingFisher Apex with 96 DW head, Extended Warranty Package |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
5400940
|
Thermo Scientific™
5400940 |
KingFisher Apex with 24 Combi head |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
A46942PKG
|
Thermo Scientific™
A46942 |
KingFisher Apex with 24 Combi Head, Extended Warranty Package |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Specifications
Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | |
>98% with 2.8 μm particle size | |
+4°C above ambient temperature up to +100°C | |
22 in. | |
18 in. | |
123 lb. | |
31 in. |
8 | |
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | |
ca. >1 μm | |
56 cm | |
46 cm | |
56 kg | |
79 cm |