missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
For chemical permeation and jet liquid protection
Supplier: Kimberly-Clark Professional™ 45647
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
- Zipper front with taped storm flap
- Interior has comfortable, cloth-like feel against the skin
- Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
4X-Large | |
White | |
Elastic | |
Attached | |
Taped | |
Hooded (respirator-fit), zipper front with storm flap, elastic wrist and ankle, exterior lamination; Saranex™ 23-P Film Laminated to Durable Fabric | |
1.48 lb. |
Saranex 23-P Fabric | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Chemicals | |
Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection | |
0.67 kg |