Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
For chemical permeation and jet liquid protection
$527.85 - $748.56
Specifications
Material Saranex 23-P Fabric
Color White
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper with Storm Flap
View More Specs
Products 10
Catalog Number Mfr. No. Size Hood Type Ankle Style Seam Type Price Quantity  
Catalog Number Mfr. No. Size Hood Type Ankle Style Seam Type Price Quantity  
29-633-925
View Documents
Kimberly-Clark Professional™
45644
X-Large Elastic Elastic Taped
Case of 12 Each for $527.85
29-633-932
View Documents
Kimberly-Clark Professional™
45666
3X-Large Attached Attached Boots Taped
Case of 10 Each for $686.53
29-633-926
View Documents
Kimberly-Clark Professional™
45645
2X-Large Elastic Elastic Taped
Case of 12 Each for $527.85
29-633-931
View Documents
Kimberly-Clark Professional™
45665
2X-Large Attached Attached Boots Taped
Case of 12 Each for $710.56
29-633-924
View Documents
Kimberly-Clark Professional™
45643
Large Attached Elastic Taped
Case of 12 Each for $527.85
29-633-930
View Documents
Kimberly-Clark Professional™
45664
X-Large Attached Attached Boots Taped
Case of 12 Each for $710.56
19-057-449E
View Documents
Kimberly-Clark Professional™
45647
4X-Large Attached Elastic Taped
Case of 10 Each for $547.03
29-633-927
View Documents
Kimberly-Clark Professional™
45646
3X-Large Elastic Elastic Taped
Case of 10 Each for $547.03
29-633-933
View Documents
Kimberly-Clark Professional™
45667
4X-Large Attached Attached Boots Taped
Case of 10 Each for $748.56
29-633-929
View Documents
Kimberly-Clark Professional™
45663
Large Attached Attached Boots Taped
Case of 12 Each for $710.56
Description
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
  • Zipper front with taped storm flap
  • Interior has comfortable, cloth-like feel against the skin
  • Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
Specifications
Saranex 23-P Fabric
Elastic
Front Zipper with Storm Flap
Chemicals
White
Hooded
Taped
Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'