Koehler™ Instrument Portable Heated Oil Test Centrifuge Tubes
Portable heated oil test centrifuge tubes
Supplier:  Koehler™ Instrument K61107
 View more versions of this product
Catalog No. 13-990-308
Explore available promotions
Description
Description
  • Centrifuge Tube, Short, 100mL, 6 in., marked in 200 parts every 4 parts above 20mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in mL every 2mL above 10mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in 200 parts (ASTM D96)
Specifications
Specifications
100 mL
Marked in mL every 2 mL above 10 mL
Heated Oil Test Centrifuge Tubes