missing translation for 'onlineSavingsMsg'
Learn More

Koehler™ Instrument Portable Heated Oil Test Centrifuge Tubes

Portable heated oil test centrifuge tubes

$365.83 - $408.23

Specifications

Capacity (Metric) 100 mL
Product Type Heated Oil Test Centrifuge Tubes
View More Specs

Products 4
Catalog Number Mfr. No. Graduations Price Quantity  
Catalog Number Mfr. No. Graduations Price Quantity  
13-990-306
View Documents
Koehler™ Instrument
K61102
Marked in 200 parts every 4 parts above 20 mL
Each for $408.23
Only null left
 
13-990-308
View Documents
Koehler™ Instrument
K61107
Marked in mL every 2 mL above 10 mL
Each for $387.57
Only null left
 
13-990-307
View Documents
Koehler™ Instrument
K61105
Marked in mL
Each for $365.83
Only null left
 
13-990-310
View Documents
Koehler™ Instrument
K61108
Marked in 200 parts
Each for $365.83
Only null left
 
Description

Description

  • Centrifuge Tube, Short, 100mL, 6 in., marked in 200 parts every 4 parts above 20mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in mL every 2mL above 10mL (ASTM D96)
  • Centrifuge Tube, Short, 100mL, 6 in., marked in 200 parts (ASTM D96)

Specifications

Specifications

100 mL
Heated Oil Test Centrifuge Tubes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'