DWK Life Sciences Kimble™ Kontes™ Microflex™ Microscale Kit with standard taper 14/10 Connection
Components have threaded standard taper 14/10 connections for fast assembly
Supplier:  DWK Life Sciences 7462200000
 View more versions of this product
Catalog No. K746220-0000
Explore available promotions
Description
Description
For experiments described in Organic Laboratory Techniques: A Microscale Approach (Saunders College Publishing, Troy, MO) and Microscale Organic Laboratory (John Wiley & Sons, New York). The Standard Kit (Part No. K746000-0000) is required to perform all experiments in those texts. Kits feature high-temperature-resistant blue nylon connectors ideal for use in temperatures to 200°C (392°F).
Order Info
In 10L x 13.5W x 2.5 in.H (25.4 x 34 x 6cm) storage case with foam insert. Catalog numbers for all kit components are listed in the accompanying chart; call your Fisher Scientific Customer Service Representative to order replacement parts.
Specifications
Specifications
Threaded Basic 14/10 Kit
2.5 in.
63.5 mm
342.9 mm
254 mm
USP Type I and ASTM E438 Type I, Class A
Claisen adapter, connecting adapter, thermometer adapter, jacketed condenser, air condenser, Hickman distillation head, Craig recrystallization tube, Craig recrystallization plug, drying tube, pear shape flask, 5mL, thin-wall, round bottom flask
13.5 in.
10 in.