DWK Life Sciences Kimble™ Kontes™ Soxhlet/Dean-Stark Apparatus with Allihn Condenser
Avoid analyte loss and sample contamination
Supplier:  DWK Life Sciences 5851500023
 View more versions of this product
Catalog No. K585150-0023
Explore available promotions
Requires : Complete assembly requires Cat. No. K586500-0023 or -0233 fritted glass extraction thimble (order separately online).
Description
Description
  • Combines Soxhlet extraction and Dean-Stark azeotropic distillation into a single process for the removal of water, chlorinated dibenzo-p-dioxins (CDDs), and chlorinated dibenzofurans (CDFs) from the same wet particulate sample
  • Does not require desiccation prior to Soxhlet extraction, so loss of analytes and sample contamination are avoided. Complete assembly requires Cat. No. K586500-0023 or -0233 fritted glass extraction thimble (order separately online)
Specifications
Specifications
Extraction Apparatus
Allihn condenser
Avoid analyte loss and sample contamination