DWK Life Sciences Kimble™ Kontes™ Soxhlet/Dean-Stark Apparatus with Allihn Condenser
Avoid analyte loss and sample contamination
$1112.24 - $3011.41
Specifications
For Use With (Application) Avoid analyte loss and sample contamination
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
K585151-0035
View Documents Promotion Details
DWK Life Sciences
5851510035
Dean/Stark Receiver
Each for $1,112.24
K585150-0023
View Documents
DWK Life Sciences
5851500023
Extraction Apparatus
Each for $3,011.41
Description
Description
  • Combines Soxhlet extraction and Dean-Stark azeotropic distillation into a single process for the removal of water, chlorinated dibenzo-p-dioxins (CDDs), and chlorinated dibenzofurans (CDFs) from the same wet particulate sample
  • Does not require desiccation prior to Soxhlet extraction, so loss of analytes and sample contamination are avoided. Complete assembly requires Cat. No. K586500-0023 or -0233 fritted glass extraction thimble (order separately online)
Specifications
Specifications
Avoid analyte loss and sample contamination
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'