missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1392406

 View more versions of this product

Catalog No. AAL1392406


Explore available promotions

Description

Description

L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in 1N HCl (50 mg/ml) and water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

L-2-Aminoadipic acid
∽206°C (decomposition)
MFCD00002636
1724348
Soluble in 1N HCl (50mg/ml) and water (slightly).
+25° (c=2 in 5N HCl)
(2S)-2-aminohexanedioic acid
92136
161.16
1118-90-7
C6H11NO4
5 g
l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
OYIFNHCXNCRBQI-BYPYZUCNSA-N
N[C@@H](CCCC(O)=O)C(O)=O
161.16
CHEBI:37023
98%

Chemical Identifiers

1118-90-7
161.16
OYIFNHCXNCRBQI-BYPYZUCNSA-N
92136
(2S)-2-aminohexanedioic acid
C6H11NO4
MFCD00002636
l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
CHEBI:37023
N[C@@H](CCCC(O)=O)C(O)=O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only