missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals
$159.85 - $528.37
Chemical Identifiers
CAS | 1118-90-7 |
---|---|
Molecular Formula | C6H11NO4 |
Molecular Weight (g/mol) | 161.16 |
MDL Number | MFCD00002636 |
InChI Key | OYIFNHCXNCRBQI-BYPYZUCNSA-N |
Synonym | l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid |
PubChem CID | 92136 |
ChEBI | CHEBI:37023 |
IUPAC Name | (2S)-2-aminohexanedioic acid |
SMILES | N[C@@H](CCCC(O)=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1392403
|
Thermo Scientific Chemicals
L1392403 |
1 g |
Each for $159.85
|
|
|||||
AAL1392406
|
Thermo Scientific Chemicals
L1392406 |
5 g |
Each for $528.37
|
|
|||||
Description
L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in 1N HCl (50 mg/ml) and water (slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
1118-90-7 | |
161.16 | |
OYIFNHCXNCRBQI-BYPYZUCNSA-N | |
92136 | |
(2S)-2-aminohexanedioic acid |
C6H11NO4 | |
MFCD00002636 | |
l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid | |
CHEBI:37023 | |
N[C@@H](CCCC(O)=O)C(O)=O |
Specifications
1118-90-7 | |
C6H11NO4 | |
1 g | |
l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid | |
OYIFNHCXNCRBQI-BYPYZUCNSA-N | |
N[C@@H](CCCC(O)=O)C(O)=O | |
161.16 | |
CHEBI:37023 | |
98% |
∽206°C (decomposition) | |
MFCD00002636 | |
1724348 | |
Soluble in 1N HCl (50mg/ml) and water (slightly). | |
+25° (c=2 in 5N HCl) | |
(2S)-2-aminohexanedioic acid | |
92136 | |
161.16 | |
L-2-Aminoadipic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only