missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-2-Octanol, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 164590010

 View more versions of this product

Catalog No. AC164590010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L(-)-2-Octanol
5978-70-1
175.0°C
Authentic
Glass bottle
1.4250 to 1.4270
MFCD00064284
01, IV, 1770
15, 6841
− 9.50
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether
CCCCCCC(C)O
130.23
CHEBI:37871
99+%
99+%
0.8300g/mL
60°C
99% min. (GC)
C8H18O
CH3(CH2)5CH(OH)CH3
1g
0.83
−9.50° (20°C neat)
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
SJWFXCIHNDVPSH-MRVPVSSYSA-N
(2R)-octan-2-ol
80080
130.23

Chemical Identifiers

5978-70-1
130.23
SJWFXCIHNDVPSH-MRVPVSSYSA-N
80080
(2R)-octan-2-ol
C8H18O
MFCD00064284
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
CHEBI:37871
CCCCCCC(C)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 227-777-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only