Learn More
L(-)-2-Octanol, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 5978-70-1 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.23 |
MDL Number | MFCD00064284 |
InChI Key | SJWFXCIHNDVPSH-MRVPVSSYSA-N |
Synonym | r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol |
PubChem CID | 80080 |
ChEBI | CHEBI:37871 |
IUPAC Name | (2R)-octan-2-ol |
SMILES | CCCCCCC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC164590010
|
Thermo Scientific Chemicals
164590010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC164590050
|
Thermo Scientific Chemicals
164590050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5978-70-1 | |
130.23 | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
80080 | |
(2R)-octan-2-ol |
C8H18O | |
MFCD00064284 | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
CHEBI:37871 | |
CCCCCCC(C)O |
Specifications
5978-70-1 | |
175.0°C | |
Authentic | |
Glass bottle | |
1.4250 to 1.4270 | |
MFCD00064284 | |
01, IV, 1770 | |
15, 6841 | |
− 9.50 | |
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether | |
CCCCCCC(C)O | |
130.23 | |
CHEBI:37871 | |
99+% |
0.8300g/mL | |
60°C | |
99% min. (GC) | |
C8H18O | |
CH3(CH2)5CH(OH)CH3 | |
1g | |
0.83 | |
−9.50° (20°C neat) | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
(2R)-octan-2-ol | |
80080 | |
130.23 | |
L(-)-2-Octanol, 99+% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 227-777-0
TSCA : TSCA
RUO â Research Use Only