missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals
$55.70 - $55.70
Chemical Identifiers
CAS | 20312-36-1 |
---|---|
Molecular Formula | C9H9O3 |
Molecular Weight (g/mol) | 165.17 |
MDL Number | MFCD00004244 |
InChI Key | VOXXWSYKYCBWHO-QMMMGPOBSA-M |
Synonym | l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid |
PubChem CID | 444718 |
ChEBI | CHEBI:43065 |
IUPAC Name | (2S)-2-hydroxy-3-phenylpropanoic acid |
SMILES | O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC150920010
|
Thermo Scientific Chemicals
150920010 |
1 g | Glass bottle |
Each for $55.70
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20312-36-1 | |
165.17 | |
VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
444718 | |
(2S)-2-hydroxy-3-phenylpropanoic acid |
C9H9O3 | |
MFCD00004244 | |
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
CHEBI:43065 | |
O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Specifications
20312-36-1 | |
Beige-Brown to White | |
98% | |
C9H9O3 | |
MFCD00004244 | |
−18.50° (24°C c=1,CH3COOH) | |
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
(2S)-2-hydroxy-3-phenylpropanoic acid | |
444718 | |
166.18 | |
Crystals, Powder or Flakes |
121°C to 125°C | |
Authentic | |
Glass bottle | |
C6H5CH2CH(OH)CO2H | |
1 g | |
-18.5 | |
Solubility in water: soluble | |
O[C@@H](CC1=CC=CC=C1)C([O-])=O | |
165.17 | |
CHEBI:43065 | |
98% | |
L(-)-3-Phenyllactic acid |
Safety and Handling
EINECSNumber : 243-726-5
RUO – Research Use Only