Learn More
L-Adrenaline, 98+%, Thermo Scientific Chemicals
$144.81 - $527.77
Chemical Identifiers
CAS | 51-43-4 |
---|---|
Molecular Formula | C9H13NO3 |
Molecular Weight (g/mol) | 183.207 |
MDL Number | MFCD00002204 |
InChI Key | UCTWMZQNUQWSLP-VIFPVBQESA-N |
Synonym | epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin |
PubChem CID | 5816 |
ChEBI | CHEBI:28918 |
IUPAC Name | 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
SMILES | CNCC(C1=CC(=C(C=C1)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0491106
|
Thermo Scientific Chemicals
L0491106 |
5 g |
Each for $144.81
|
|
||||||
AAL0491114
|
Thermo Scientific Chemicals
L0491114 |
25 g |
Each for $527.77
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-43-4 | |
183.207 | |
UCTWMZQNUQWSLP-VIFPVBQESA-N | |
5816 | |
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
C9H13NO3 | |
MFCD00002204 | |
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin | |
CHEBI:28918 | |
CNCC(C1=CC(=C(C=C1)O)O)O |
Specifications
51-43-4 | |
C9H13NO3 | |
5 g | |
2368277 | |
14,3619 | |
UCTWMZQNUQWSLP-VIFPVBQESA-N | |
CNCC(C1=CC(=C(C=C1)O)O)O | |
183.207 | |
CHEBI:28918 | |
≥98% |
216°C to 219°C | |
MFCD00002204 | |
UN2811 | |
Air and light sensitive | |
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin | |
−52° (c=1 in 0.6N HCl) | |
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
5816 | |
183.2 | |
L-Adrenaline |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P314-P330-P361-P501c
H301+H331-H310-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-098-7
RTECSNumber : DO2625000
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only