missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6339303

 View more versions of this product

Catalog No. AAH6339303

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Alanine 4-nitroanilide hydrochloride
Yellow
MFCD00039088
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl
1 g
2802426
98%
31796-55-1
C9H12ClN3O3
6303731
YEXRLSXNWLNHQR-RGMNGODLSA-N
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride
245.663
245.66
Powder

Chemical Identifiers

31796-55-1
245.663
YEXRLSXNWLNHQR-RGMNGODLSA-N
2802426
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl
C9H12ClN3O3
MFCD00039088
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 250-811-0

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only